Learn More
Cayman Chemical Abl Substrate Peptide (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502919316
Encompass_Preferred
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro
Catalog No. 50-291-9316 Supplier Cayman Chemical Supplier No. 369691 MG
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro