Learn More
Cayman Chemical Abl Substrate Peptide (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502915494
Encompass_Preferred
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro
Catalog No. 50-291-5494 Supplier Cayman Chemical Supplier No. 369695 MG
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro