Learn More
Cayman Chemical Abl Substrate Peptide (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502919319
Encompass_Preferred
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro
Catalog No. 50-291-9319 Supplier Cayman Chemical Supplier No. 3696910 MG
A peptide substrate for Abl; selectively phosphorylated by Abl over c-Src kinase at 20 µM; has been used to quantify Abl kinase activity in vitro