Learn More
Sino Biological Recombinant Human TPX2 Protein (Full Length, His Tag)
Catalog No. 502572724
Encompass_Preferred
Recombinant full length human TPX2 protein was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. This product can be used for Aurora A activation in vitro.
Catalog No. 50-257-2724 Supplier Sino Biological Supplier No. T4030H1000
Recombinant full length human TPX2 protein was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. This product can be used for Aurora A activation in vitro.