Learn More
Enzo Life Sciences Abl1 (human), (recombinant) (His-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5620005
Encompass_Preferred
Alternative name: Abelson murine leukemia viral oncogene homolog 1. Purity: ≥70% (SDS-PAGE). Formulation: Liquid. In 50mM sodium phosphate, pH 7.0, containing 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2094