Learn More
Sino Biological Recombinant Human c-Abl/ABL1 Protein (Q252H, GST Tag), Active
Catalog No. 502576086
Encompass_Preferred
Recombinant human ABL1 (Q252H) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-6086 Supplier Sino Biological Supplier No. A0312FG500
Recombinant human ABL1 (Q252H) (27-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.