Learn More
Enzo Life Sciences Abl1 (mouse), (recombinant) (His-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5630005
Encompass_Preferred
Alternative name: Abelson murine leukemia viral oncogene homolog 1. Concentration: 0.1mg/ml. Purity: ≥70% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 150mM sodium chloride, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF and 25% glycerol. Source: Produced in insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2095