Learn More
Sino Biological Recombinant Human BTK Protein (Full Length, R562W, GST Tag)
Catalog No. 502575765
Encompass_Preferred
Recombinant full length human BTK (R562W) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5765 Supplier Sino Biological Supplier No. B1012JG20
Recombinant full length human BTK (R562W) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.