Learn More
Sino Biological Recombinant Human ADCK3 Protein (GST Tag), Active
Catalog No. 502575876
Encompass_Preferred
Recombinant human ADCK3 (156-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5876 Supplier Sino Biological Supplier No. A37112G500
Recombinant human ADCK3 (156-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.