Learn More
Radiometer America Radiometer ABL80 CO-OX Solution Pack
SDP
Catalog No. NC1873403
Encompass_Preferred
ABL80 CO-OX Solution Pack
Solution pack for Radiometer ABL80 CO-OX blood gas analyzer.
ABL80 FLEX Blood Gas Analyzer
Test results when and where you need them. The ABL80 FLEX CO-OX analyzer provides blood gas, electrolytes, glucose and CO-oximetry parameters with all the advantages of a compact system, including simplicity and reliability.
This item is not returnable. View return policy
Catalog No. NC1873403 Supplier Radiometer America Supplier No. 944252
ABL80 CO-OX Solution Pack
Solution pack for Radiometer ABL80 CO-OX blood gas analyzer.
ABL80 FLEX Blood Gas Analyzer
Test results when and where you need them. The ABL80 FLEX CO-OX analyzer provides blood gas, electrolytes, glucose and CO-oximetry parameters with all the advantages of a compact system, including simplicity and reliability.