Learn More
Agilent Technologies ABLE K COMPETENT CELLS
Catalog No. 50125028
Encompass
ABLE K Electroporation-Competent Cells reduces the number of common cloning vectors by 10-fold and enhance the probability that a toxic clone will be propagated. ABLE K strains are alternatives to time-consuming subcloning projects
Catalog No. 50-125-028 Supplier Agilent Technologies Supplier No. 200172
ABLE K Electroporation-Competent Cells reduces the number of common cloning vectors by 10-fold and enhance the probability that a toxic clone will be propagated. ABLE K strains are alternatives to time-consuming subcloning projects