Learn More
Reprocell ABLE MAG STIR BASE 30ML/100ML
Catalog No. NC1813136
Encompass
ABLE Bioreactor Magnetic Stir System Base 30 mL and 100 mL; Six position stir base for 30 mL and 100 mL flasksThis Stir System Base works with the Able Bioreactor System Controller and Motor (Cat. No. ABBWDW-1013) to provide consistent, heat-free stirring of 30 mL and 100 mL disposable bioreactors. REPROCELL mfg# ABBWBP03NoS-6
Catalog No. NC1813136 Supplier Reprocell Supplier No. ABBWBP03N0S6
ABLE Bioreactor Magnetic Stir System Base 30 mL and 100 mL; Six position stir base for 30 mL and 100 mL flasksThis Stir System Base works with the Able Bioreactor System Controller and Motor (Cat. No. ABBWDW-1013) to provide consistent, heat-free stirring of 30 mL and 100 mL disposable bioreactors. REPROCELL mfg# ABBWBP03NoS-6