Learn More
Sino Biological Peptide Substrates: Abltide
Catalog No. 502569890
Encompass_Preferred
The Abltide peptide sequence (EAIYAAPFAKKK) is based on the C-terminus of Abl.
Catalog No. 50-256-9890 Supplier Sino Biological Supplier No. A02581000
The Abltide peptide sequence (EAIYAAPFAKKK) is based on the C-terminus of Abl.