Learn More
CPC Scientific 5TAMRA-Lys-Lys-Gly-Glu-Ala-Ile-Tyr-Ala-Ala-Pro-Phe-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110230
Encompass
TAMRA-labeled Abltide (Ab/Em = 544/572 nm) that is a peptide substrate for Abl kinase (Abl protein tyrosine kinase), a partner in the gag-Abl fusion protein of the Abelson murine leukemia virus. Used in Western blot and kinase assays. ONE-LETTER SEQUENCE: 5TAMRA-KKGEAIYAAPFA-NH2MOLECULAR FORMULA: C85H113N17O19MOLECULAR WEIGHT:1677STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Reddy, PE. et al. Proc. Natl. Acad. Sci. USA 80 (1983)
Catalog No. 50-211-0230 Supplier CPC Scientific Supplier No. PROK007B
TAMRA-labeled Abltide (Ab/Em = 544/572 nm) that is a peptide substrate for Abl kinase (Abl protein tyrosine kinase), a partner in the gag-Abl fusion protein of the Abelson murine leukemia virus. Used in Western blot and kinase assays. ONE-LETTER SEQUENCE: 5TAMRA-KKGEAIYAAPFA-NH2MOLECULAR FORMULA: C85H113N17O19MOLECULAR WEIGHT:1677STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Reddy, PE. et al. Proc. Natl. Acad. Sci. USA 80 (1983)