Learn More
Lonza Walkersville ABM Astrocyte Basal Medium
Catalog No. NC1948115
Encompass
Clonetics AGM Astrocyte Growth Medium has been developed to support the growth of primary human astrocytes.Clonetics Media Products are provided as convenient BulletKits which contain basal medium and necessary growth factors. Basal medium and SingleQuots Kit growth factors are available separately.Clonetics Media Prodcuts are specially designed to support the growth of human and animal primary cells. Media systems have been specifically developed and optimized for various cell systems.Media are supplied in several formats:Basal Medium - contains no growth factors cytokines, or supplementsSingleQuots - growth factors, cytokines, and supplementsBulletKits - basal media and SingleQuots Kit packaged togetherContent:Astrocyte basal medium (500 mL) (no growth factors)
Catalog No. NC1948115 Supplier Lonza Walkersville Supplier No. CC3187
Clonetics AGM Astrocyte Growth Medium has been developed to support the growth of primary human astrocytes.Clonetics Media Products are provided as convenient BulletKits which contain basal medium and necessary growth factors. Basal medium and SingleQuots Kit growth factors are available separately.Clonetics Media Prodcuts are specially designed to support the growth of human and animal primary cells. Media systems have been specifically developed and optimized for various cell systems.Media are supplied in several formats:Basal Medium - contains no growth factors cytokines, or supplementsSingleQuots - growth factors, cytokines, and supplementsBulletKits - basal media and SingleQuots Kit packaged togetherContent:Astrocyte basal medium (500 mL) (no growth factors)