Learn More
Medchemexpress LLC HY-124801 10mg Medchemexpress, ABMA CAS:332108-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12480110MG
Encompass_Preferred
Medchemexpress, HY-124801 10mg ABMA CAS:332108-65-3 ABMA is a broad-spectrum inhibitor of intracellular toxins and pathogens. ABMA efficiently protects cells against various toxins and pathogens including viruses, intracellular bacteria and parasite. ABMA selectively acts at host cell late endosomes rather than targeting toxin or pathogen itself. ABMA has broad-spectrum anti-infection activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0011
 Restricted Item