Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 10p Subtelomere (Cy5) FISH Probe

Catalog No. 89029274 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-029-274 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-029-274 Supplier Abnova™ Supplier No. FE0095
Add to Cart
Add to Cart

Includes

FISH Hybridization Buffer (80μL)

Labeled FISH probes for identification of subtelomere aberrations using Fluorescent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye Cy5
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Includes FISH Hybridization Buffer (80 uL)
Location 10p Subtelomere
Product Type 10p Subtelomere(Cy5) FISH Probe
Quantity 20 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.