Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 11-dehydro-TXB2 ELISA Kit

Catalog No. 89100186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-186 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-186 Supplier Abnova™ Supplier No. KA0314
Add to Cart
Add to Cart

Includes

  • Goat anti-Rabbit IgG Microtiter Plate
  • 11-dehydro-TXB2 ELISA Conjugate
  • 11-dehydro-TXB2 ELISA Antibody: A yellow solution of a rabbit polyclonal antibody to 11-dehydro-TXB2
  • Assay Buffer: Tris buffered saline, containing proteins and sodium azide as preservative
  • Wash Buffer Concentrate: Tris buffered saline containing detergents
  • 11-dehydro-Thromboxane B2 Standard: A solution of 100,000pg/mL 11-dehydro-TXB2
  • pNpp Substrate: A solution of p-nitrophenyl phosphate in buffer. Ready to use
  • Stop Solution: A solution of trisodium phosphate in water. Keep tightly capped. Caution: Caustic
  • Plate Sealer

Use to measure quantitatively 11-dehydro-Thromboxane B2 in tissue culture media and urine. A competitive immunoassay.

The kit for the quantitative measurement of 11-dehydro-Thromboxane B2 uses a polyclonal antibody to 11-dehydro-TXB2 to bind, in a competitive manner, the 11-dehydro-TXB2 in the sample or an alkaline phosphatase molecule which has 11-dehydro-TXB2 covalently attached to it

Specifications

Calibration Range 9.8 to 10,000pg/mL
Conjugate AP
Description 11-dehydro-TXB2 ELISA Kit is a competitive immunoassay for the quantitative determination of 11-dehydro-Thromboxane B2 in tissue culture media and urine.
Detection Limit 4.31pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Tissue Culture Media, Urine
Sample Volume 100 μL
Sensitivity 4.31 pg/mL
Packaging 96-well
Content And Storage The Standard and Conjugate must be stored at -20°C; Others store at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type 11-dehydro-TXB2 ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.