Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 12(S)-HETE ELISA Kit

Catalog No. 89100165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-165 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-165 Supplier Abnova™ Supplier No. KA0306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay Buffer: Tris buffered saline containing proteins and sodium azide as preservative
  • 12(S)-HETE Standard: A solution of 500,000pg/mL 12(S)-HETE
  • Goat anti-Rabbit IgG Microtiter Plate: A clear plate of break-apart strips coated with a goat anti-rabbit polyclonal antibody
  • 12(S)-HETE Antibody: A yellow solution of a rabbit polyclonal antibody to 12(S)-HETE
  • 12(S)-HETE Conjugate: A blue solution of 12(S)-HETE conjugated to alkaline phosphatase
  • Wash Buffer Concentrate: Tris buffered saline containing detergents
  • pNpp Substrate: A solution of p-nitrophenyl phosphate
  • Stop Solution
  • Plate Sealer

Use to measure quantitatively 12(S)-HETE in biological fluids. A competitive immunoassay.

Specifications

Calibration Range 195 to 50,000pg/mL
Conjugate AP
Description 12(S)-HETE ELISA Kit is a competitive immunoassay for the quantitative determination of 12(S)-HETE in biological fluids.
Detection Limit 146pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma
Sample Volume 100 μL
Sensitivity 146.3 pg/mL
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type 12(S)-HETE ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.