Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 15-deoxy-Delta12,14-PGJ2 ELISA Kit

Catalog No. 89100168 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-168 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-168 Supplier Abnova™ Supplier No. KA0303
Only null left
Add to Cart
Add to Cart

Includes

  • Goat anti-Rabbit IgG Microtiter Plate, 96 Wells
  • 15-d-PGJ2 ELISA Conjugate, 5mL
  • 15-d-PGJ2 ELISA Antibody, 5mL
  • Assay Buffer, 27mL
  • Wash Buffer Concentrate, 27mL
  • 15-deoxy-▴12,14-Prostaglandin J2
  • Standard, 0.5mL
  • p-Npp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 2 slices

Use to measure quantitatively 15-deoxy-Delta12,14-Prostaglandin J2 (15-d-PGJ2) in biological fluids. A competitive immunoassay.

The kit uses a polyclonal antibody to 15-d-PGJ2 to bind, in a competitive manner, the 15-d-PGJ2 in the sample or an alkaline phosphatase molecule which has 15-d-PGJ2 covalently attached to it

Specifications

Calibration Range 195 to 200,000pg/mL
Conjugate AP
Description 15-deoxy-Delta12,14-PGJ2 ELISA Kit is a competitive immunoassay for the quantitative determination of 15-deoxy-Delta12,14-Prostaglandin J2 (15-d-PGJ2) in biological fluids.
Detection Limit 36.8pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Saliva, Tissue culture media and Urine
Sample Volume 100 μL
Sensitivity 36.8 pg/mL
Packaging 96-well
Content And Storage Store all component at 4°C, except standard and conjugate at -20°C
For Use With (Application) Quantification
Product Type 15-deoxy-Delta12,14-PGJ2 ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.