Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 15(S)-HETE ELISA Kit

Catalog No. 89100164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-164 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-164 Supplier Abnova™ Supplier No. KA0307
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Goat anti-Rabbit IgG Microtiter Plate
  • 15(S)-HETE EIA Conjugate: A blue solution of alkaline phosphatase conjugated with 15(S)-HETE
  • 15(S)-HETE EIA Antibody: A yellow solution of a rabbit polyclonal antibody to 15(S)-HETE
  • Assay Buffer: Tris buffered saline containing proteins and sodium azide as preservative
  • Wash Buffer Concentrate: Tris buffered saline containing detergents
  • 15(S)-HETE Standard: A solution of 200,000pg/mL 15(S)-HETE
  • pNpp Substrate: A solution of p-nitrophenyl phosphate in buffer. Ready to use
  • Stop Solution: A solution of trisodium phosphate in water. Keep tightly capped. Caution: Caustic
  • Plate Sealers

Use to measure quantitatively 15(S)-HETE in biological fluids. A competitive immunoassay.

The kit uses a polyclonal antibody to 15(S)-HETE to bind, in a competitive manner, the 15(S)-HETE in the standard or sample or an alkaline phosphatase molecule which has 15(S)-HETE covalently attached to it

Specifications

Calibration Range 78.1 to 20,000pg/mL
Conjugate AP
Description 15(S)-HETE ELISA Kit is a competitive immunoassay for the quantitative determination of 15(S)-HETE in biological fluids.
Detection Limit 69.21pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 100 μL
Sensitivity 69.21 pg/mL
Packaging 96-well
Content And Storage The standard and conjugate must be stored at -20°C; others store at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type 15(S)-HETE ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.