Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 17-OH-Progesterone ELISA Kit

Catalog No. 89028712 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-712 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-712 Supplier Abnova™ Supplier No. KA1896
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of active free 17-OH-Progesterone in Saliva

Specifications

Conjugate HRP
Description 17-OH-Progesterone ELISA Kit is an enzyme immunoassay for the quantitative measurement of active free 17-OH-Progesterone in saliva.
Detection Limit 3.6pg/mL
Detection Method Colorimetric
Includes 96-well microtiter coated (12 x 8 strip) with Anti-17a-OHP Antibody (Polyclonal), 26mL Enzyme Conjugate, 1mL Cortisol standard A, 1mL Cortisol standard B, 1mL Cortisol standard C, 1mL Cortisol standard D, 1mL Cortisol standard E,, 1mL Cortisol standard F, 1.0mL/vial, 2 vials Control 1 and 2, 30mL wash solution (40X concentrated), 25mL Substrate solution 14mL stopping solutions
Quantity 1 Kit
Sample Type Saliva
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
Form Liquid
For Use With (Application) Quantification
Product Type 17-OH-Progesterone ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.