Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 2,4D ELISA Kit

Catalog No. 89024531 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-531 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-531 Supplier Abnova™ Supplier No. KA1411
 Restricted Item
Add to Cart
Add to Cart

Competitive Binding Enzyme Immunoassay for the quantitative measurement of 2,4-Dichlorophenoxyacetic Acid

Specifications

Conjugate AP
Description 2,4D ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of 2,4-dichlorophenoxyacetic acid.
Detection Method Colorimetric
Includes 12 Strip of 8 detachable well, 96-well microtiter plate, 0.25mL positive calibrator, 10.5mL 2,4D-Alkaline Phosphatase Conjugate (24D-ALP)), 10.5mL p-Nitrophenyl Phosphate (pNPP) Substrate, 15mL Wash Buffer, 6mL stop solution
Quantity 1 Kit
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type 2,4D ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.