Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 5-hydroxyindoleacetic acid ELISA Kit

Catalog No. 89028614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-614 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-614 Supplier Abnova™ Supplier No. KA1881
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative determination of 5-HIAA

Specifications

Calibration Range 2.4 to 24.3mg/L
Description 5-hydroxyindoleacetic acid ELISA Kit is Enzyme immunoassay for the quantitative determination of 5-HIAA (5-hydroxyindoleacetic acid).
Detection Limit 0.17mg/L
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, 1 x 96-well reaction plate, 1 x 4 adhesive foil, 1 x 20mL Wash Buffer concentrate, 1 x 12mL Enzyme Conjugate, 1 x 22mL Diluent, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 4mL standard A, 1 x 4mL standard B, 1 x 4mL standard C,
Quantity 1 Kit
Sample Type Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type 5-Hydroxyindoleacetic Acid ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.