Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 5a-Androstane-3a, 17b-Diol Glucuronide ELISA Kit

Catalog No. 89028713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-713 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-713 Supplier Abnova™ Supplier No. KA1897
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of 5a-Androstane-3a, 17b-Diol Glucuronide in Human Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description 5a-Androstane-3a, 17b-Diol Glucuronide ELISA Kit is an enzyme immunoassay for the quantitative measurement of 5a-Androstane-3a, 17b-Diol Glucuronide in human serum.
Detection Limit 0.1ng/mL
Detection Method Colorimetric
Includes 1 x 96-well Antibody coated microwell plate, 1 x 300μL HRP Conjugate, 1 x 2mL calibrator A, 0.6mL/each calibrator B to F, 0.6mL/ each Control 1 and 2, 1 x 12mL Substrate, 1 x 12mL stopping solution,, 1 x 50mL Wash Buffer concentrate, 1 x
Quantity 1 Kit
Sample Type Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type 5a-Androstane-3a, 17b-Diol Glucuronide ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.