Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ 6-keto-PGF1-alpha ELISA Kit

Catalog No. 89100174 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-174 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-174 Supplier Abnova™ Supplier No. KA0295
Add to Cart
Add to Cart

Includes

  • Donkey anti-Sheep, IgG Microtiter Plate, 96 wells/plate
  • 6-keto-PGF1α ELISA Conjugate, 6mL
  • 6-keto-PGF1α ELISA Antibody, 6mL
  • Assay Buffer, 30mL
  • Wash Buffer Concentrate, 30mL
  • 6-keto-Prostaglandin F1α Standard, 0.5mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 slice

Use to measure quantitatively 6-keto-PGF1-alpha in biological fluids. A competitive immunoassay.

The kit uses a polyclonal antibody to 6-keto-PGF1α to bind, in a competitive manner, the 6-keto-PGF1α in the standards or sample or an alkaline phosphatase molecule which has 6-keto-PGF1α covalently attached to it

Specifications

Binding Capacity The kit uses a polyclonal antibody to 6-keto-PGF1α to bind, in a competitive manner, the 6-keto-PGF1α in the standards or sample or an alkaline phosphatase molecule which has 6-keto-PGF1α covalently attached to it.
Calibration Range 3.2 to 50,000pg/mL
Conjugate AP
Description 6-keto-PGF1-alpha ELISA Kit is a competitive immunoassay for the quantitative determination of 6-keto-PGF1-alpha in biological fluids.
Detection Limit 1.40pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Saliva, Serum, Tissue culture media, Urine
Sample Volume 100 μL
Sensitivity 1.4 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type 6-keto-PGF1-alpha ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.