Learn More
Description
Includes
- 8-OHdG Microtiter Plate: Precoated With 8-OHdG (8 x12 wells, Split Type), 1 Plate
- Primary Antibody: Monoclonal Antibody Specific For 8-OHdG
- Primary Antibody Dilution Buffer: Phosphate Buffered Saline
- Secondary Antibody: HRP-Conjugated Antibody
- Secondary Antibody Dilution Buffer: Phosphate Buffered Saline
- Chromogen: 3,3',5,5'-Tetramethylbenzidine
- Chromogen Dilution Buffer: Hydrogen Peroxide/Citrate-Phosphate Buffered Saline
- Washing Buffer (5x): Concentrated Phosphate Buffered Saline, 2 at 26mL
- Stop Solution: 1M Phosphoric Acid
- Standards 1-6
- Plate Seal: Adhesive sheet for covering plate
Specifications
Specifications
| Calibration Range | 0.5 to 200ng/mL |
| Conjugate | HRP |
| Description | 8-hydroxy-2'-deoxyguanosine ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of 8-hydroxy-2'-deoxyguanosine. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Sample Type | Plasma, Serum, Tissue, Urine |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.