Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ A2M (Human) ELISA Kit

Catalog No. p-4392015 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Volume:
25 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
89-022-914 25 μL
89-100-236 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-022-914 Supplier Abnova™ Supplier No. KA0487
Only null left
Add to Cart
Add to Cart

Includes

  • Polystyrene microplate coated with polyclonal antibody against α - 2 - Macroglobulin, 96 (8 x 12) wells
  • Pre - cut, pressure - sensitive sealing tapes, which can be cut to fit the format of the individual assay, 3 in no.
  • Human α - 2 - Macroglobulin in a buffered protein base (lyophilized), 20μg
  • Biotinylated α-2-Macroglobulin, lyophilized, 1 vial
  • 10-fold EIA diluent concentrated buffered protein base, 30mL
  • 20-fold concentrated wash buffered surfactant, 30mL
  • 100-fold Streptavidin-Peroxidase conjugate (SP conjugate), 80μL
  • Ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • 0.5N hydrochloric acid to stop the chromogen substrate reaction, 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human A2M in Plasma and Serum

Specifications

Calibration Range 1.25 to 40μg/mL
Conjugate Biotin
Cross Reactivity Human
Description A2M (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human A2M in plasma and serum.
Detection Limit 1μg/mL
Detection Method Colorimetric
Includes 96-well Polystyrene microplate coated with Polyclonal Antibody, precut, pressure sensitive sealing tapes, 20μg Human a-2-macroglobulin in a Buffered protein base, 1 vial Biotinylated a-2-Macroglobulin, lyophilized, 30mL 10-fold EIA Dilue
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type A2m (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.