Learn More
Specifications
Specifications
| Calibration Range | 0.313 to 100ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Mouse, Rat |
| Description | A2m (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse A2m in cell culture supernatants. |
| Detection Limit | 0.3ng/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 20ng Mouse A2m standard, 1 vial Biotinylated A2m Antibody (100X), 30mL MIX Diluent concentrate (10X), 30mL Wash Buffer 20X, 90 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL Chromogen Substrate, 12mL stop solution |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum, Urine |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.