Learn More
Abnova™ Acetyl-Histone H4 ChIP Kit
Catalog No. 89027029 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-029 1 Kit
1 options
Catalog No. 89-027-029 Supplier Abnova™ Supplier No. KA0670
Add to Cart
Add to Cart
Acetyl-Histone H4 ChIP Kit is for investigating the interactions of histone H4 acetylation and DNA in vivo efficiently.
  • The fastest procedure available, which can be finished within 5 hours
  • Strip microplate format makes the assay flexible; manual or high throughput
  • Columns for DNA purification are included; save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
Specifications
Description Acetyl-Histone H4 ChIP Kit is for investigating the interactions of histone H4 acetylation and DNA in vivo efficiently.
Content And Storage Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, acetyl-histone H4 antibody, proteinase K, 8 well strips. Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Chromatin Immunoprecipitation
Type ChIP Kit