Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ACTH (Human) ELISA Kit

Catalog No. 89024457 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-457 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-457 Supplier Abnova™ Supplier No. KA0916
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Microwells coated with Streptavidin, 12 x 8 x 1
  • Biotinylated ACTH antibody (reagent 1), 2.7mL
  • Peroxidase (enzyme) labeled ACTH antibody, 2.7mL
  • Wash concentrate, 30mL
  • TMB substrate, 15mL
  • Stop solution, 20mL
  • Calibrators, 2mL x 5
  • Zero calibrator, 4mL
  • Controls 1 & 2 (CTRL), 2mL

For quantitative determination of ACTH in Human Plasma

Specifications

Conjugate Biotin
Cross Reactivity Human
Description ACTH (Human) ELISA Kit is used for the quantitative determination of ACTH (Adrenocorticotropic Hormone) in human plasma.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 12 x 8 x 1 Microwell coated with Streptavidin, 2.7mL Biotinylated ACTH Antibody, 2.7mL Peroxidase labeled ACTH Antibody, 30mL Wash concentrate, 15mL TMB Substrate, 20mL stop solution, 2mL x 5 calibrator, 4mL zero calibrator, 2mL controls 1 and 2
Quantity 96 Assays
Sample Type Plasma
Sample Volume 200 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ACTH (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.