Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ACTH (Mouse/Rat) ELISA Kit

Catalog No. 89024458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-458 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-458 Supplier Abnova™ Supplier No. KA0917
Add to Cart
Add to Cart

Ultra sensitive method (Less than 1 pg/mL) used for the quantitative determination of ACTH (Adrenocorticotropic Hormone) in mouse/rat plasma

  • Sample Volume: 200μL
  • Sample Type: Plasma
  • Calibration Range: 5 to 700pg/mL
  • Conjugate: Biotin

Quantification

Specifications

Conjugate Biotin
Cross Reactivity Mouse, Rat
Description ACTH (Mouse/Rat) ELISA Kit (Chemiluninescence) is an ultra sensitive method (Less than 1 pg/mL) used for the quantitative determination of ACTH (Adrenocorticotropic Hormone) in mouse/rat plasma.
Detection Method Colorimetric
Includes 6 x 2 x 8 Microwell coated with Streptavidin, 4mL ACTH standard zero: 1 bottle, ready to use, 2mL ACTH standard: 5 bottles (lyophilized), 2.7mL Biotinylated ACTH Antibody (Reagent 1), 2.7mL Enzyme labeled ACTH Antibody (Reagent 2), 4mL luminol Substrate,
Quantity 96 Assays
Sample Type Plasma
Sample Volume 200 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ACTH (Mouse/Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.