Learn More
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Mouse |
| Description | Activin A (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Activin A. |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant A Activin standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-A activin Antibody, 12mL Antibody Diluent Buffer, 130μL avidin-biotin-per |
| Quantity | 1 Kit |
| Sample Type | Body Fluid, Cell Culture Supernatant, Serum, Tissue Lysate |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at -20°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.