Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ADAM17 (Human) ELISA Kit

Catalog No. 89100425 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-425 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-425 Supplier Abnova™ Supplier No. KA2145

Includes

  • TACE Microplate (Item A): 96 wells coated with anti-Human TACE
  • Wash Buffer Concentrate (20x) (Item B)
  • Standard Protein (Item C): Human TACE. 1 vial is enough to run each standard in duplicate
  • Assay Diluent C (Item L): Diluent buffer
  • Assay Diluent B (Item E): 5x concentrated buffer
  • Detection Antibody TACE (Item F): Biotinylated anti-Human TACE. Each vial is enough to assay half the microplate
  • HRP-Streptavidin Concentrate (Item G): 200x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H): 3,3',5,5'-tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively human ADAM17. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human TACE coated on a 96-well plate

Specifications

Calibration Range 78.15 to 5000pg/mL
Conjugate HRP
Cross Reactivity Human
Description ADAM17 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human ADAM17.
Detection Limit 70pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity 70 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ADAM17 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.