Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ADIPOQ (Human) ELISA Kit

Catalog No. 89100726 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Milk, Plasma, Saliva, Serum, Urine
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
Plasma (Citrate, EDTA, Heparin), Serum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-726 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-028-041 CCS, Milk, Plasma, Saliva, Serum, Urine
89-100-012 CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine
89-100-013 Plasma (Citrate, EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-726 Supplier Abnova™ Supplier No. KA3019
Add to Cart
Add to Cart

For detection of ADIPOQ in Human Urine and Murine, Adiponectin in Plasma, Serum and cell culture supernatants

Specifications

Calibration Range 1.56 to 100ng/mL
Conjugate Biotin
Cross Reactivity Human
Description ADIPOQ (Human) ELISA Kit is intended for the quantitative measurement of human ADIPOQ.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human Adiponectin antibody, Lyophilized Human Adiponectin standards (100ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human Adiponectin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) (130uL), ABC diluent buffer (12mL),TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X)(30mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type ADIPOQ (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.