Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Adipoq (Mouse) ELISA Kit

Catalog No. 89028165 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Urine
CCS, Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-165 CCS, Plasma, Serum, Urine
89-100-011 CCS, Serum
89-100-727 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-028-165 Supplier Abnova™ Supplier No. KA1025
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Adipoq in Urine, Plasma, Serum and cell culture supernatants

Specifications

Calibration Range 0.78 to 50ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description Adipoq (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse adipoq in urine, plasma, serum, and cell culture supernatants.
Detection Limit 1ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 125ng Mouse adiponectin standard, 1 vial Biotinylated adiponectin Antibody (100X), 30mL mix diluent concentrate (10X), 30mL Wash Buffer 20X, 90 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ADIPOQ (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.