Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Adipoq (Mouse) ELISA Kit

Catalog No. 89100727 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Urine
CCS, Serum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-727 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-028-165 CCS, Plasma, Serum, Urine
89-100-011 CCS, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-727 Supplier Abnova™ Supplier No. KA3020
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Adipoq in Urine, Plasma, Serum and cell culture supernatants

Specifications

Calibration Range 312 to 20000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Adipoq (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Adipoq.
Detection Limit 10 pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Mouse Adiponectin antibody, Lyophilized Adiponectin standards(10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti -Mouse Adiponectin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), 25X Wash buffer (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Adipoq (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.