Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Aflatoxin B ELISA Kit

Catalog No. 89024532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-532 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-532 Supplier Abnova™ Supplier No. KA1412
Add to Cart
Add to Cart

Includes

  • 96-wells microtiter plate (No. S), Twelve strips of 8 detachable wells, coated with rabbit Anti-Aflatoxin antibody, 1 in no.
  • Calibrator containing 0, 0.5, 2.0 and 8.0ng/mL of Aflatoxin B1, 0.6mL, 4 vials
  • (No. 3) Aflatoxin-Alkaline Phosphatase conjugate (AFX-ALP), 10.5mL
  • (No. 5) p-Nitrophenyl Phosphate (pNPP) substrate. Ready to use, 10.5mL
  • (No. 6) Wash Buffer (10xPBS-Tween). Dilute 10 fold with distilled or deionized water to 150mL prior to use, 15mL
  • (No. 7) Stop Solution, 3N NaOH, 6mL

Competitive Binding Enzyme Immunoassay for the quantitative measurement of Aflatoxin B

Specifications

Conjugate AP
Description Aflatoxin B ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of aflatoxin B.
Detection Method Colorimetric
Includes 12 Strip of 8 detachable well, 96-well microtiter plate, 1.2mL negative calibrator, 3 vials positive calibrator, 10.5mL aflatoxin-alkaline Phosphatase Conjugate, 10.5mL p-nitrophenyl Phosphate (pNPP)Substrate, 15mL Wash Buffer, 6mL stop solution
Quantity 1 Kit
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Aflatoxin B ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.