Learn More
Description
AFP-L3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human AFP-L3
Quantification
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Includes | 1 Assay plate, standard 2, 20mL sample diluent, 10mL assay diluent A, 10mL assay diluent B, 120μL detection reagent A, 120μL detection reagent B, 30mL 25X Wash Buffer, 10mL Substrate, 10mL stop solution, 5mL plate seale |
| Quantity | 96 Assays |
| Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Homogenate |
| Sample Volume | 100 μL |
| Content And Storage | Kit at -20°C and substrate at 4°C |
| For Use With (Application) | Quantification, for the quantitative measurement of Human AFP-L3 |
| Product Type | AFP-L3 (Human) ELISA kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.