Learn More
Description
Includes
- Lyophilized recombinant human Ager standard, 10ng/tube x 2
- One 96-well plate precoated with anti- human Ager antibody, 96 wells
- Sample diluent buffer, 30mL
- Biotinylated anti-human Ager antibody: dilution 1:100, 130μL
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Description | AGER (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human AGER. |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell plate, 10ng x 2 vials lyophilized recombinant Human AGER standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human AGER Antibody, 12mL Antibody Diluent Buffer, 130μL avidin-bi |
| Quantity | 1 Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at -20°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.