Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ AHSG (Human) ELISA Kit

Catalog No. p-4392086 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum

Sandwich Enzyme Immunoassay for the quantitative measurement of Human AHSG

Specifications

Calibration Range 0.312 to 20ng/mL
Conjugate Biotin
Cross Reactivity Human
Description Human AHSG ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human AHSG.
Detection Limit <10 pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell plate, 10ng x 2 vials lyophilized recombinant Human AGER standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human AGER Antibody, 12mL Antibody Diluent Buffer, 130μL avidin-bi
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type AHSG (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.