Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ AKT1 (pS473)/Pan AKT1 (Human/Mouse/Rat) ELISA Kit

Catalog No. 89100432 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-432 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-432 Supplier Abnova™ Supplier No. KA2152

Includes

  • Akt Microplate: 96 wells coated with anti-pan-Akt antibody
  • Wash Buffer Concentrate (20x)
  • Assay Diluent
  • Detection Antibody Akt (Ser473): rabbit anti-phospho-Akt (Ser473)
  • Detection Antibody Akt: rabbit anti-pan-Akt
  • HRP-conjugated Anti-rabbit IgG, 25μL of 1,000x HRP-conjugated Anti-rabbit IgG concentrate
  • TMB One-Step Substrate Reagent: 12mL of 3,3',5,5'-tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution: 2M sulfuric acid
  • Cell Lysate Buffer: 5mL 2x cell lysis buffer (not including protease and phosphatase inhibitors)
  • Positive Control A431S003-1: 2 vial of lyophilized powder from A431 cell lysate

Use to detect qualitatively human, mouse, rat phospho AKT1 (S473) and Pan AKT1. Enzyme-linked immunosorbent assay.

AKT1 (pS473)/Pan AKT1 (Human/Mouse/Rat) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates

Specifications

Conjugate HRP
Cross Reactivity Human, Mouse, Rat
Description AKT1 (pS473)/Pan AKT1 (Human/Mouse/Rat) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human, mouse, rat phospho AKT1 (S473) and Pan AKT1.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Qualitative
Product Type AKT1 (pS473)/Pan AKT1 (Human/Mouse/Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.