Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Rabbit) ELISA Kit

Catalog No. 89100370 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-370 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-370 Supplier Abnova™ Supplier No. KA2112
Add to Cart
Add to Cart

Includes

  • Rabbit Albumin Microplate: A 96-well polystyrene microplate coated with a polyclonal chicken antibody against rabbit albumin
  • Sealing Tapes
  • Rabbit Albumin Standard: Rabbit albumin in a buffered protein base
  • Biotinylated Albumin
  • MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL)
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (80μL)
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL)
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively rabbit albumin. A competitive enzyme immunoassay.

The Albumin (Rabbit) ELISA Kit employs a quantitative competitive enzyme immunoassay technique that measures rabbit plasma, serum, cell culture supernatant and urine albumin in less than 3 hours

Specifications

Conjugate Biotin
Cross Reactivity Rabbit
Description Albumin (Rabbit) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rabbit albumin.
Detection Limit 0.19μg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Albumin (Rabbit) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.