Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Rat), Biotin, ELISA Kit

Catalog No. 89100222 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Urine
Plasma, serum and urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-222 CCS, Urine
89-100-223 Plasma, serum and urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-222 Supplier Abnova™ Supplier No. KA0502
Add to Cart
Add to Cart

Includes

  • antibody coated microwells 96 wells
  • Rat Albumin Standard 800ng
  • Biotinylated Albumin antibody (100x) 1 vial
  • MIX Diluent Concentrate (10x) 30mL
  • Wash Buffer Concentrate (20x) 30mL x 2
  • Streptavidin-Peroxidase Conjugate (SP Conjugate) 80
  • Chromogen Substrate 8mL
  • Stop Solution: 12mL

For quantitative measurement of rat Nppb.

Specifications

Calibration Range 3 to 50ng/mL
Conjugate Biotin
Cross Reactivity Rat
Description Albumin (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat albumin in cell culture supernatant and urine.
Detection Limit 0.7 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Albumin (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.