Learn More
Specifications
Specifications
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Description | ALCAM (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human ALCAM. |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP-streptavidian concentrate, 12mL TMB one-step Substrate reagent, 8mL stop solut |
| Quantity | 96 Assays |
| Sample Type | Cell Culture Supernatant, Plasma, Serum, Urine |
| Sample Volume | 100 μL |
| Content And Storage | Store at 4°C. Standard (recombinant protein) should be stored at -20°C or -80°C (recommended at -80°C) after reconstitution. The kit for long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.