Learn More
Description
- Sample Type: Plasma, Serum, Urine
- Sample Volume: 50μL
- Detection Range: 10pg/mL
- Detection Method: Colorimetric
Quantification
Specifications
Specifications
| Description | Aldosterone ELISA Kit is an enzyme immunoassay for the quantitative determination of Aldosterone in human serum, plasma, and urine. |
| Detection Method | Colorimetric |
| Includes | 1 Coated microtiter plate, 1 x 300μL Conjugate concentrate, 15mL Assay Buffer, 50mL Wash Buffer concentrate, 16mL Substrate, 6mL stopping solution, 1 x 2mL standard A, 1 x 0,6mL standard B, 1 x 0,6mL standard C, 1 x 0,6mL standard D, 1 x |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum, Urine |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | Aldosterone ELISA kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.