Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alpha-1-Acid Glycoprotein (Chicken) ELISA Kit

Catalog No. 89100619 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-619 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-619 Supplier Abnova™ Supplier No. KA2423
Add to Cart
Add to Cart

Includes

  • Anti-chicken α-1-AGP antibody coated microtiter plate, 96 wells (12 detachable strips x 8)
  • Enzyme Conjugate Reagent ,11mL
  • Reference standard (lyophilized), 1 vial
  • 10x Diluent, 25mL
  • 20x Wash Solution, 50mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively Alpha-1-Acid Glycoprotein in serum.

The Alpha-1-Acid Glycoprotein (Chicken) is based on a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Conjugate HRP
Cross Reactivity Chicken
Description Alpha-1-Acid Glycoprotein (Chicken) ELISA Kit is for the quantitative measurement of Alpha-1-Acid Glycoprotein in serum.
Detection Limit 2.3 to 150ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Alpha-1-Acid Glycoprotein (Chicken) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.