Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alpha 1-anti-Chymotrypsin (Human) ELISA Kit

Catalog No. 89100354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-354 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-354 Supplier Abnova™ Supplier No. KA2128
Add to Cart
Add to Cart

Includes

  • Diluent Concentrate (Running Buffer)
  • Wash Solution Concentrate
  • Enzyme-Antibody Conjugate 100X: affinity purified anti-Human Alpha 1-Antichymotrypsin antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti-Human Alpha 1-Antichymotrypsin ELISA Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame. Each well is coated with affinity purified anti-Human CYT
  • Human Alpha 1-Antichymotrypsin Calibrator: a lyophilized Human Alpha 1-Antichymotrypsin calibrator

Measure human Alpha 1-Anti-Chymotrypsin. Two-site enzyme linked immunoassay.

In this assay the Alpha 1-Antichymotrypsin present in samples reacts with the anti-Alpha 1-Antichymotrypsin antibodies which have been adsorbed to the surface of polystyrene microtitre wells

Specifications

Calibration Range 6.25 to 400ng/mL
Conjugate HRP
Cross Reactivity Human
Description Alpha 1-Anti-Chymotrypsin (Human) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of human Alpha 1-Anti-Chymotrypsin.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Alpha 1-Anti-Chymotrypsin (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.