Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Alpha1-Microglobulin (Human) ELISA Kit

Catalog No. 89024526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-526 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-526 Supplier Abnova™ Supplier No. KA1406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sandwich Enzyme Immunoassay for quantitative measurement of Human Alpha1-microglobulin in Plasma, Serum, Urine, Saliva, Milk and cell culture supernatant

Specifications

Conjugate Biotin
Cross Reactivity Human
Description AMBP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human alpha1-microglobulin in plasma, serum, urine, saliva, milk, and cell culture supernatant.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 160ng Human alpha1-microglobulin standard, 80μL Biotinylated 1M Antibody (100X), 30mL EIA Diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80μL Streptavidin-Peroxidase Conjugate (S
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
Form Lyophilized
For Use With (Application) Quantification
Product Type AMBP (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.