Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ANA Profile ELISA Kit

Catalog No. 89027217 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-217 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-217 Supplier Abnova™ Supplier No. KA1079
Add to Cart
Add to Cart

Qualitative, for the determination ANA autoantibodies directed against RNP-70, RNP/Sm, Sm, SS-A, SS-B, Scl-70, Centromer B and Jo-1 in Human Serum or Plasma

Specifications

Conjugate HRP
Description ANA Profile ELISA Kit is an indirect solid phase ELISA for the determination ANA autoantibodies directed against RNP-70, RNP/Sm, Sm, SS-A, SS-B, Scl-70, Centromer B and Jo-1 in human serum or plasma.
Detection Method Colorimetric
Includes 96-well microplate coated with highly purified Antigens RNP-70 (row A), RNP/Sm (row B), Sm (row C), SS-A (row D), SS-B (row E), Scl-70 (row F), Centromer B (row G), Jo-1 (row H), 2 2, 5mL Anti-ANA controls, 20mL Sample Buffer(5x), 15mL Enzyme Conjugate solution vial, 15mL TMB Substrate solution vial, 15mL stop solution vial, 20mL Wash solution(50X)
Quantity 96 Tests
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ANA Profile ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.